2'-(Allyloxy)-6'-hydroxyacetophenon
Lieferant: Apollo Scientific
|
Achtung
|
OR22017-250MG
APOSOR22017-250MGEA
116
EUR
APOSOR22017-250MG
2'-(Allyloxy)-6'-hydroxyacetophenon
2'-(Allyloxy)-6'-hydroxyacetophenon
Formel:
C₁₁H₁₂O₃ Schmelzpunkt: 45,5…46.5 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
23226-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...