1-(5-Amino-2-hydroxyphenyl)ethanone

Lieferant: Apollo Scientific

OR22048-1G
APOSOR22048-1GEA 223 EUR
APOSOR22048-1G
1-(5-Amino-2-hydroxyphenyl)ethanone
1-(5-Amino-2-hydroxyphenyl)ethanone
Formel: C₈H₉NO₂
Molecular Weight: 151,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00100397
CAS-Nummer: 50-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR