Bis(4-hydroxyphenyl)sulfid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4,4'-Thiodiphenol
OR22056-500G
APOSOR22056-500GEA
84.1
EUR
APOSOR22056-500G
Bis(4-hydroxyphenyl)sulfid 98%
Bis(4-hydroxyphenyl)sulfid
Formel:
C₁₂H₁₀O₂S Molecular Weight: 218,28 g/mol Schmelzpunkt: 151…156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002349 CAS-Nummer: 2664-63-3 EINECS: 220-197-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...