5-Aminobenzene-1,3-diol hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR22059-250MG
APOSOR22059-250MGEA
27.7
EUR
APOSOR22059-250MG
5-Aminobenzene-1,3-diol hydrochloride
5-Aminobenzene-1,3-diol hydrochloride
Formel:
C₆H₈ClNO₂ Molecular Weight: 161,5862 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00662897 CAS-Nummer: 6318-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...