Ethyl 6-iodochromone-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 6-iodo-4-oxo-4H-1-benzopyran-2-carboxylate, Ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate

OR22061-250MG
APOSOR22061-250MGEA 21.9 EUR
APOSOR22061-250MG
Ethyl 6-iodochromone-2-carboxylate
Ethyl 6-iodochromone-2-carboxylate
Formel: C₁₂H₉IO₄
Molecular Weight: 344,102 g/mol
Schmelzpunkt: 148…150 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00100378
CAS-Nummer: 35204-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR