4-Hexylanilin
Lieferant: Apollo Scientific
|
Achtung
|
OR22069-1G
APOSOR22069-1GEA
21.9
EUR
APOSOR22069-1G
4-Hexylanilin
4-Hexylanilin
Formel:
C₁₂H₁₉N Molecular Weight: 177,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007927 CAS-Nummer: 33228-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...