3-(4-Ethoxybenzoyl)propionsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Ethoxyphenyl)-4-oxobutanoic acid

OR22072-1G
APOSOR22072-1GEA 26.1 EUR
APOSOR22072-1G
3-(4-Ethoxybenzoyl)propionsäure
3-(4-Ethoxybenzoyl)propionsäure
Formel: C₁₂H₁₄O₄
Molecular Weight: 222,24 g/mol
Schmelzpunkt: 136…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00100659
CAS-Nummer: 53623-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR