3-Brom-5-ethoxy-4-hydroxybenzaldehyd

Lieferant: Apollo Scientific
Achtung

OR22077-1G
APOSOR22077-1GEA 21.9 EUR
APOSOR22077-1G
3-Brom-5-ethoxy-4-hydroxybenzaldehyd
3-Bromo-5-ethoxy-4-hydroxybenzaldehyde
Formel: C₉H₉BrO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00173904
CAS-Nummer: 3111-37-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR