4-Benzyl 2-ethyl 3-(2-ethoxy-2-oxoethyl)-5-methyl-1H-pyrrole-2,4-dicarboxylate

Lieferant: Apollo Scientific

OR22109-1G
APOSOR22109-1GEA 183 EUR
APOSOR22109-1G
4-Benzyl 2-ethyl 3-(2-ethoxy-2-oxoethyl)-5-methyl-1H-pyrrole-2,4-dicarboxylate
4-Benzyl 2-ethyl 3-(2-ethoxy-2-oxoethyl)-5-methyl-1H-pyrrole-2,4-dicarboxylate
Formel: C₂₀H₂₃NO₆
Molecular Weight: 373,4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00102152
CAS-Nummer: 52887-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR