Ethyl-3-(3-nitrophenyl)-3-oxopropanoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3-(3-nitrophenyl)-3-oxopropanoate

OR22122-5G
APOSOR22122-5GEA 72.5 EUR
APOSOR22122-5G
Ethyl-3-(3-nitrophenyl)-3-oxopropanoate
Ethyl-3-(3-nitrophenyl)-3-oxopropanoate
Formel: C₁₁H₁₁NO₅
Molecular Weight: 237,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00126483
CAS-Nummer: 52119-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR