2,6-Dibromonitrobenzene 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR2213-1G
APOSOR2213-1GEA
21.9
EUR
APOSOR2213-1G
2,6-Dibromonitrobenzene 95+%
2,6-Dibromonitrobenzene
Formel:
C₆H₃Br₂NO₂ Schmelzpunkt: 78…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09998726 CAS-Nummer: 13402-32-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...