2-Amino-6-ethylpyridin 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 6-Ethylpyridin-2-amin, 6-Ethyl-2-pyridylamin

OR22212-1G
APOSOR22212-1GEA 21.9 EUR
APOSOR22212-1G
2-Amino-6-ethylpyridin 95%
2-Amino-6-ethylpyridine
Formel: C₇H₁₀N₂
Molecular Weight: 122.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00218265
CAS-Nummer: 21717-29-3
EINECS: 244-540-7
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR