2-(2-Diethylamino)ethylpyridine

Lieferant: Apollo Scientific

Synonyme: N,N-Diethyl-2-pyridin-2-ylethylamine

OR22219-1G
APOSOR22219-1GEA 72.5 EUR
APOSOR22219-1G
2-(2-Diethylamino)ethylpyridine
2-(2-Diethylamino)ethylpyridine
Formel: C₁₁H₁₈N₂
Molecular Weight: 178,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014633
CAS-Nummer: 25877-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR