3-Methyl-N-[(1E)-3-(4-methylphenyl)-3-oxoprop-1-en-1-yl]pyridinium chloride

Lieferant: Apollo Scientific
Achtung

Synonyme: N-[(1E)-3-(4-Methylphenyl)-3-oxoprop-1-en-1-yl]-3-picolinium chloride

OR22248-500MG
APOSOR22248-500MGEA 29 EUR
APOSOR22248-500MG
3-Methyl-N-[(1E)-3-(4-methylphenyl)-3-oxoprop-1-en-1-yl]pyridinium chloride
3-Methyl-N-[(1E)-3-(4-methylphenyl)-3-oxoprop-1-en-1-yl]pyridinium chloride
Formel: C₁₆H₁₆NO·Cl
Molecular Weight: 273,757 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00204658
CAS-Nummer: 19518-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR