1-(4-Methoxyphenyl)-3-(3-methylpyridinium-1-yl)prop-2-en-1-one chloride

Lieferant: Apollo Scientific
Achtung

Synonyme: N-[(1E)-3-(4-Methoxyphenyl)-3-oxoprop-1-en-1-yl]-3-methylpyridinium chloride

OR22249-500MG
APOSOR22249-500MGEA 29 EUR
APOSOR22249-500MG
1-(4-Methoxyphenyl)-3-(3-methylpyridinium-1-yl)prop-2-en-1-one chloride
1-(4-Methoxyphenyl)-3-(3-methylpyridinium-1-yl)prop-2-en-1-one chloride
Formel: C₁₆H₁₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00204660
CAS-Nummer: 1080601-76-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR