N-[(1E)-3-(4-Chlorophenyl)-3-oxoprop-1-en-1-yl]-3-methylpyridinium chloride

Lieferant: Apollo Scientific
Achtung

Synonyme: N-[(1E)-3-(4-Chlorophenyl)-3-oxoprop-1-en-1-yl]-3-picolinium chloride

OR22250-500MG
APOSOR22250-500MGEA 29 EUR
APOSOR22250-500MG
N-[(1E)-3-(4-Chlorophenyl)-3-oxoprop-1-en-1-yl]-3-methylpyridinium chloride
N-[(1E)-3-(4-Chlorophenyl)-3-oxoprop-1-en-1-yl]-3-methylpyridinium chloride
Formel: C₁₅H₁₃ClNO·Cl
Molecular Weight: 294,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00204661
CAS-Nummer: 19518-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR