3-exo-Aminobicyclo[2.2.1]heptane-2-exo-carboxylic acid, N-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Boc-3-exo-aminobicyclo[2.2.1]heptane-2-exo-carboxylic acid
OR2227-1G
APOSOR2227-1GEA
383
EUR
APOSOR2227-1G
3-exo-Aminobicyclo[2.2.1]heptane-2-exo-carboxylic acid, N-BOC protected
3-exo-Aminobicyclo[2.2.1]heptane-2-exo-carboxylic acid, N-BOC protected
Formel:
C₁₃H₂₁NO₄ Molecular Weight: 255,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682613 CAS-Nummer: 1212373-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...