Hexylmercaptoacetat

Lieferant: Apollo Scientific
Achtung

Synonyme: Hex-1-yl sulphanylacetate

OR22277-1G
APOSOR22277-1GEA 92.8 EUR
APOSOR22277-1G
Hexylmercaptoacetat
Hexylmercaptoacetat
Formel: C₈H₁₆O₂S
Molecular Weight: 176.28 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 20292-01-7
EINECS: 243-696-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR