2-Aminobenzimidazol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzimidazol-2-ylamin, 2-Amino-1H-benzimidazol
OR22305-100G
OR22305-25G
OR22305-500G
APOSOR22305-100GEA
55.1
EUR
APOSOR22305-100G
APOSOR22305-25G
APOSOR22305-500G
2-Aminobenzimidazol 97%
2-Aminobenzimidazole
Formel:
C₇H₇N₃ Molecular Weight: 133.15 g/mol Siedepunkt: 381 Schmelzpunkt: 228…232 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005596 CAS-Nummer: 934-32-7 EINECS: 213-280-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...