1-Methyl-1H-indole-3-carbaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR22346-100G
APOSOR22346-100GEA
159
EUR
APOSOR22346-100G
1-Methyl-1H-indole-3-carbaldehyde 98%
1-Methyl-1H-indole-3-carbaldehyde
Formel:
C₁₀H₉NO Schmelzpunkt: 68…70 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00014570 CAS-Nummer: 19012-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...