(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected 97%
Lieferant: Apollo Scientific
Synonyme:
(2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-((tert-butoxycarbonyl)amino)pyrrolidine-2-carboxylic acid, Fmoc-trans-4-N-Boc-Pro-OH
OR2235-250MG
APOSOR2235-250MGEA
364
EUR
APOSOR2235-250MG
(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected 97%
(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected
Formel:
C₂₅H₂₈N₂O₆ Molecular Weight: 452,5 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01860707 CAS-Nummer: 273222-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...