4-Chloro-1-methyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR2244-1G
APOSOR2244-1GEA 52.2 EUR
APOSOR2244-1G
4-Chloro-1-methyl-1H-indazole
4-Chloro-1-methyl-1H-indazole
Formel: C₈H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09998749
CAS-Nummer: 162502-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR