4-(1-Adamantyl)phenol 97%
Lieferant: Apollo Scientific
OR22443-5G
APOSOR22443-5GEA
0
EUR
APOSOR22443-5G
4-(1-Adamantyl)phenol 97%
4-(1-Adamantyl)phenol
Formel:
C₁₆H₂₀O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00168143 CAS-Nummer: 29799-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...