Methyl-4-aminobutanoate hydrochloride
Lieferant: Apollo Scientific
Synonyme:
H-γ-Abu-OMe.HCl
OR22446-100G
APOSOR22446-100GEA
58
EUR
APOSOR22446-100G
Methyl-4-aminobutanoate hydrochloride
Methyl-4-aminobutanoate hydrochloride
Formel:
C₅H₁₁NO₂·HCl Molecular Weight: 153.61 g/mol Schmelzpunkt: 120…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043270 CAS-Nummer: 13031-60-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...