1-[4-(1-Adamantyl)phenoxy]-3-piperidin-1-ylpropan-2-ol hydrochloride

Lieferant: Apollo Scientific

OR22453-1G
APOSOR22453-1GEA 342 EUR
APOSOR22453-1G
1-[4-(1-Adamantyl)phenoxy]-3-piperidin-1-ylpropan-2-ol hydrochloride
1-[4-(1-Adamantyl)phenoxy]-3-piperidin-1-ylpropan-2-ol hydrochloride
Formel: C₂₄H₃₅NO₂·ClH
Molecular Weight: 406,001 g/mol
Schmelzpunkt: 249…252 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00218803
CAS-Nummer: 175136-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR