4-Nitro-2-picolin-N-oxid
Lieferant: Apollo Scientific
|
Achtung
|
OR22458-5G
APOSOR22458-5GEA
21.9
EUR
APOSOR22458-5G
4-Nitro-2-picolin-N-oxid
4-Nitro-2-picolin-N-oxid
Formel:
C₆H₆N₂O₃ Schmelzpunkt: 155…159 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014625 CAS-Nummer: 5470-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...