1-(1-Adamantyl)ethylamine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Rimantadine Hydrochloride, alpha-Methyl-1-adamantanemethylamine Hydrochloride
OR22496-10G
APOSOR22496-10GEA
114
EUR
APOSOR22496-10G
1-(1-Adamantyl)ethylamine hydrochloride
1-(1-Adamantyl)ethylamine hydrochloride
Formel:
C₁₂H₂₁N Molecular Weight: 215,77 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00072023 CAS-Nummer: 1501-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...