1-(N-Boc)-3-cyanoazetidine

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-cyanoazetidine-1-carboxylate, Azetidine-3-carbonitrile, N-BOC protected

OR2250-25G
APOSOR2250-25GEA 112 EUR
APOSOR2250-25G
1-(N-Boc)-3-cyanoazetidine
1-(N-Boc)-3-cyanoazetidine
Formel: C₉H₁₄N₂O₂
Molecular Weight: 182,22 g/mol
Schmelzpunkt: 70…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06796640
CAS-Nummer: 142253-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR