3-Amino-6-chlor-1-methyl-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR2265-1G
APOSOR2265-1GEA 197 EUR
APOSOR2265-1G
3-Amino-6-chlor-1-methyl-1H-indazol
3-Amino-6-chloro-1-methyl-1H-indazole
Formel: C₈H₈ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09991761
CAS-Nummer: 1031927-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR