Methylpicolinat 96%
Lieferant: Apollo Scientific
Synonyme:
Methyl pyridine-2-carboxylate
OR22699-100G
APOSOR22699-100GEA
104
EUR
APOSOR22699-100G
Methylpicolinat 96%
Methylpicolinat
Formel:
C₇H₇NO₂ Molecular Weight: 137,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038038 CAS-Nummer: 2459-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...