N-Decyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-10, SB-10) 98%

Lieferant: Apollo Scientific

Synonyme: 3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulphonate, 3-(Decyldimethylazaniumyl)propane-1-sulphonate

OR2270T-25G
APOSOR2270T-25GEA 201 EUR
APOSOR2270T-25G
N-Decyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-10, SB-10) 98%
N-Decyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-10, SB-10)

A zwitterionic detergent.

Formel: C₁₅H₃₃NO₃S
Molecular Weight: 307,4924 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036908
CAS-Nummer: 15163-36-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR