2,2-Dimethylcyclopropanecarboxamide
Lieferant: Apollo Scientific
|
Achtung
|
OR22745-1G
OR22745-250MG
OR22745-500MG
OR22745-2.5G
OR22745-5G
APOSOR22745-1GEA
132
EUR
APOSOR22745-1G
APOSOR22745-250MG
APOSOR22745-500MG
APOSOR22745-2.5G
APOSOR22745-5G
2,2-Dimethylcyclopropanecarboxamide
2,2-Dimethylcyclopropanecarboxamide
Formel:
C₆H₁₁NO Schmelzpunkt: 176…177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00829406 CAS-Nummer: 1759-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...