Diethyl 2-aminomalonate, N-BOC protected 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Diethyl 2-(tert-Butoxycarbonylamino)malonate, Diethyl 2-[(tert-butoxycarbonyl)amino]malonate, Diethyl 2-[(tert-butoxycarbonyl)amino]propane-1,3-dioate
OR22746-5G
APOSOR22746-5GEA
33.5
EUR
APOSOR22746-5G
Diethyl 2-aminomalonate, N-BOC protected 97%
Diethyl 2-aminomalonate, N-BOC protected
Formel:
C₁₂H₂₁NO₆ Molecular Weight: 275,298 g/mol Siedepunkt: 218 °C Dichte: 1,079 g/cm³ (25 °C) Flammpunkt: 113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00239423 CAS-Nummer: 102831-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...