4-Cyclohexylbenzenesulphonyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR2275-1G
OR2275-500MG
OR2275-5G
APOSOR2275-1GEA
0
EUR
APOSOR2275-1G
APOSOR2275-500MG
APOSOR2275-5G
4-Cyclohexylbenzenesulphonyl chloride 97%
4-Cyclohexylbenzenesulphonyl chloride
Formel:
C₁₂H₁₅ClO₂S Schmelzpunkt: 42…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00625713 CAS-Nummer: 56354-57-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...