4-Amino-N,N-dimethylbenzolsulfonamid

Lieferant: Apollo Scientific
Achtung

Synonyme: Anilin-4-sulfonsäuredimethylamid, 4-Aminobenzolsulfonsäuredimethylamid

OR2276-250MG
APOSOR2276-250MGEA 21.9 EUR
APOSOR2276-250MG
4-Amino-N,N-dimethylbenzolsulfonamid
4-Amino-N,N-dimethylbenzolsulfonamid
Formel: C₈H₁₂N₂O₂S
Molecular Weight: 200.26 g/mol
Schmelzpunkt: 168…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031428
CAS-Nummer: 1709-59-7
EINECS: 216-970-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR