2,5-Dimethoxyterephthalaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-dimethoxyterephthalaldehyde, 2,5-Dimethoxybenzene-1,4-dicarboxaldehyde

OR22763-1G
APOSOR22763-1GEA 203 EUR
APOSOR22763-1G
2,5-Dimethoxyterephthalaldehyde
2,5-Dimethoxyterephthalaldehyde
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,184 g/mol
Schmelzpunkt: 209…213 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017606
CAS-Nummer: 7310-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR