2-Methyl-5-sulphamoylbenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Carboxy-4-methylbenzenesulphonamide, 5-(Aminosulphonyl)-2-methylbenzoic acid, 5-Sulphamoyl-o-toluic acid
OR2277-1G
APOSOR2277-1GEA
87
EUR
APOSOR2277-1G
2-Methyl-5-sulphamoylbenzoic acid
2-Methyl-5-sulphamoylbenzoic acid
Formel:
C₈H₉NO₄S Molecular Weight: 215,226 g/mol Schmelzpunkt: 243…250 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07362354 CAS-Nummer: 20532-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...