1-(5-Bromo-1H-indol-3-yl)-2-[4-(pyrimidin-2-yl)piperazin-1-yl]ethane-1,2-dione

Lieferant: Apollo Scientific
Achtung

OR22784-250MG
APOSOR22784-250MGEA 31.9 EUR
APOSOR22784-250MG
1-(5-Bromo-1H-indol-3-yl)-2-[4-(pyrimidin-2-yl)piperazin-1-yl]ethane-1,2-dione
1-(5-Bromo-1H-indol-3-yl)-2-[4-(pyrimidin-2-yl)piperazin-1-yl]ethane-1,2-dione
Formel: C₁₈H₁₆BrN₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00829501
CAS-Nummer: 256417-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR