tert-Butyl (5-methylpyrazin-2-yl)carbamate

Lieferant: Apollo Scientific

Synonyme: 2-(Boc-amino)-5-methylpyrazine

OR2282-5G
APOSOR2282-5GEA 63.8 EUR
APOSOR2282-5G
tert-Butyl (5-methylpyrazin-2-yl)carbamate
tert-Butyl (5-methylpyrazin-2-yl)carbamate
Formel: C₁₀H₁₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09907674
CAS-Nummer: 369638-68-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR