4-Acetyl-4-(ethoxycarbonyl)heptane-1,7-dioic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Acetyl-4-(ethoxycarbonyl)heptanedioic acid, 4-Acetyl-4-(ethoxycarbonyl)pimelic acid, Ethyl 2,2-bis(2-carboxyethyl)acetoacetate, 3-Acetyl-3-(ethoxycarbonyl)pentane-1,5-dicarboxylic acid

OR22879-1G
APOSOR22879-1GEA 82.8 EUR
APOSOR22879-1G
4-Acetyl-4-(ethoxycarbonyl)heptane-1,7-dioic acid
4-Acetyl-4-(ethoxycarbonyl)heptane-1,7-dioic acid
Formel: C₁₂H₁₈O₇
Molecular Weight: 274,267 g/mol
Schmelzpunkt: 105…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052373
CAS-Nummer: 72653-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR