5-(Dimethylamino)pentan-2-one

Lieferant: Apollo Scientific

OR22887-250MG
APOSOR22887-250MGEA 249 EUR
APOSOR22887-250MG
5-(Dimethylamino)pentan-2-one
5-(Dimethylamino)pentan-2-one
Formel: C₇H₁₅NO
Molecular Weight: 129.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00048233
CAS-Nummer: 43018-61-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR