2-Amino-5-(hydroxymethyl)pyrazine, 2-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[(tert-Butoxycarbonyl)amino]-5-(hydroxymethyl)pyrazine, tert-Butyl [5-(hydroxymethyl)pyrazin-2-yl]carbamate

OR2290-50MG
APOSOR2290-50MGEA 50.9 EUR
APOSOR2290-50MG
2-Amino-5-(hydroxymethyl)pyrazine, 2-BOC protected
2-Amino-5-(hydroxymethyl)pyrazine, 2-BOC protected
Formel: C₁₀H₁₅N₃O₃
Molecular Weight: 225,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09952117
CAS-Nummer: 874476-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR