Diethyl-4-oxocyclohexane-1,1-dicarboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Diethyl 4-oxocyclohexane-1,1-dicarboxylate

OR22900-1G
APOSOR22900-1GEA 0 EUR
APOSOR22900-1G
Diethyl-4-oxocyclohexane-1,1-dicarboxylate 97%
Diethyl-4-oxocyclohexane-1,1-dicarboxylate
Formel: C₁₂H₁₈O₅
Molecular Weight: 242,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00177942
CAS-Nummer: 55704-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR