Ethyl 2,4-dichlorophenylacetate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-(2,4-dichlorophenyl)ethanoate, Ethyl (2,4-dichlorophenyl)acetate, Ethyl 2-(2,4-dichlorophenyl)acetate

OR2296-500MG
APOSOR2296-500MGEA 48 EUR
APOSOR2296-500MG
Ethyl 2,4-dichlorophenylacetate 97%
Ethyl 2,4-dichlorophenylacetate
Formel: C₁₀H₁₀Cl₂O₂
Molecular Weight: 233,091 g/mol
Siedepunkt: 88 °C (0,05 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD01566538
CAS-Nummer: 41022-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR