3,4-(Methylenedioxy)thiobenzamide

Lieferant: Apollo Scientific

Synonyme: Benzo[d][1,3]dioxole-5-carbothioamide, 1,3-Benzodioxole-5-carbothioamide

OR22962-1G
APOSOR22962-1GEA 248 EUR
APOSOR22962-1G
3,4-(Methylenedioxy)thiobenzamide
3,4-(Methylenedioxy)thiobenzamide
Formel: C₈H₇NO₂S
Molecular Weight: 181,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02677739
CAS-Nummer: 15884-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR