5-(Pyridin-2-yl)thiophene-2-thiocarboxamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Carbamothioyl-5-(pyridin-2-yl)thiophene, 2-(5-Carbamothioylthien-2-yl)pyridine
OR22963-250MG
APOSOR22963-250MGEA
79.9
EUR
APOSOR22963-250MG
5-(Pyridin-2-yl)thiophene-2-thiocarboxamide
5-(Pyridin-2-yl)thiophene-2-thiocarboxamide
Formel:
C₁₀H₈N₂S₂ Molecular Weight: 220,314 g/mol Schmelzpunkt: 198…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02677740 CAS-Nummer: 306934-91-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...