benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate

OR22966-5G
APOSOR22966-5GEA 120 EUR
APOSOR22966-5G
benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate 97%
benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate
Formel: C₁₄H₁₈N₂O₃
Molecular Weight: 262.304 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01568760
CAS-Nummer: 167757-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR