2-Aminofuran, N-BOC protected 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[(tert-Butoxycarbonyl)amino]furan, 2-(Boc-amino)furan, tert-Butyl (fur-2-yl)carbamate

OR22968-1G OR22968-250MG
APOSOR22968-1GEA 60.9 EUR
APOSOR22968-1G APOSOR22968-250MG
2-Aminofuran, N-BOC protected 97%
2-Aminofuran, N-BOC protected
Formel: C₉H₁₃NO₃
Schmelzpunkt: 98…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682023
CAS-Nummer: 56267-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR