2-Chlormethyl-1,4-benzodioxan
Lieferant: Apollo Scientific
|
Achtung
|
OR22971-1G
APOSOR22971-1GEA
125
EUR
APOSOR22971-1G
2-Chlormethyl-1,4-benzodioxan
2-Chlormethyl-1,4-benzodioxan
Formel:
C₉H₉ClO₂ Siedepunkt: 96 °C/0,4mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014666 CAS-Nummer: 2164-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...