Ethyl-2-amino-4-(4-bromophenyl)thiophene-3-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-amino-4-(4-bromophenyl)thiophene-3-carboxylate

OR22974-1G
APOSOR22974-1GEA 58 EUR
APOSOR22974-1G
Ethyl-2-amino-4-(4-bromophenyl)thiophene-3-carboxylate
Ethyl-2-amino-4-(4-bromophenyl)thiophene-3-carboxylate
Formel: C₁₃H₁₂BrNO₂S
Molecular Weight: 326.22 g/mol
Schmelzpunkt: 120…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00435056
CAS-Nummer: 306934-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR